Assay Details
Gene Symbol: LOC5575216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850012.1 | XP_021705704.1 | 6 - 7 | 2739 | 66 | |
XM_021850013.1 | XP_021705705.1 | 6 - 7 | 2739 | 66 | ||
GenBank mRNA | JN030894.1 | - | 3 - 4 | 798 | 66 | |
KC439534.1 | - | 3 - 4 | 1243 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
