Assay Details
Gene Symbol: LOC5577461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663337.2 | XP_001663387.2 | 3 - 4 | 856 | 83 | |
XM_021843406.1 | XP_021699098.1 | 4 - 5 | 695 | 83 | ||
XM_021843407.1 | XP_021699099.1 | 3 - 4 | 784 | 83 | ||
XM_021843408.1 | XP_021699100.1 | 2 - 3 | 125 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021843408.1 transcripts.
Set Membership: |
![]() ![]() |
