Assay Details
Gene Symbol: LOC5577461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663337.2 | XP_001663387.2 | 13 - 14 | 4102 | 81 | |
XM_021843406.1 | XP_021699098.1 | 14 - 15 | 3941 | 81 | ||
XM_021843407.1 | XP_021699099.1 | 13 - 14 | 4030 | 81 | ||
XM_021843408.1 | XP_021699100.1 | 12 - 13 | 3371 | 81 |
