Assay Details
Gene Symbol: LOC5577461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663337.2 | XP_001663387.2 | 10 - 11 | 3503 | 93 | |
XM_021843406.1 | XP_021699098.1 | 11 - 12 | 3342 | 93 | ||
XM_021843407.1 | XP_021699099.1 | 10 - 11 | 3431 | 93 | ||
XM_021843408.1 | XP_021699100.1 | 9 - 10 | 2772 | 93 |
