Assay Details
Gene Symbol: LOC5577461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663337.2 | XP_001663387.2 | 7 - 8 | 2377 | 69 | |
XM_021843406.1 | XP_021699098.1 | 8 - 9 | 2216 | 69 | ||
XM_021843407.1 | XP_021699099.1 | 7 - 8 | 2305 | 69 | ||
XM_021843408.1 | XP_021699100.1 | 6 - 7 | 1646 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
