Assay Details
Gene Symbol: LOC5577895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842937.1 | XP_021698629.1 | 8 - 9 | 2808 | 75 | |
XM_021842938.1 | XP_021698630.1 | 8 - 9 | 2633 | 75 | ||
XM_021842939.1 | XP_021698631.1 | 8 - 9 | 2808 | 75 | ||
XM_021842940.1 | XP_021698632.1 | 8 - 9 | 2808 | 75 | ||
XM_021842942.1 | XP_021698634.1 | 8 - 9 | 2808 | 75 | ||
XM_021842943.1 | XP_021698635.1 | 8 - 9 | 2808 | 75 | ||
XM_021842944.1 | XP_021698636.1 | 8 - 9 | 2808 | 75 | ||
XM_021842945.1 | XP_021698637.1 | 8 - 9 | 2808 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
