Assay Details
Gene Symbol: LOC5574316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661298.2 | XP_001661348.1 | 11 - 12 | 5931 | 67 | |
XM_021838871.1 | XP_021694563.1 | 12 - 13 | 5938 | 67 | ||
XM_021838872.1 | XP_021694564.1 | 11 - 12 | 5930 | 67 | ||
XM_021838873.1 | XP_021694565.1 | 12 - 13 | 5940 | 67 | ||
XM_021838874.1 | XP_021694566.1 | 12 - 13 | 5893 | 67 | ||
XM_021838875.1 | XP_021694567.1 | 12 - 13 | 5890 | 67 | ||
XM_021838876.1 | XP_021694568.1 | 11 - 12 | 3705 | 67 | ||
XM_021838877.1 | XP_021694569.1 | 11 - 12 | 4108 | 67 | ||
XM_021838878.1 | XP_021694570.1 | 11 - 12 | 3696 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
