Assay Details
Gene Symbol: LOC5574316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661298.2 | XP_001661348.1 | 5 - 6 | 4837 | 76 | |
XM_021838871.1 | XP_021694563.1 | 5 - 6 | 4835 | 76 | ||
XM_021838872.1 | XP_021694564.1 | 5 - 6 | 4836 | 76 | ||
XM_021838873.1 | XP_021694565.1 | 5 - 6 | 4837 | 76 | ||
XM_021838874.1 | XP_021694566.1 | 5 - 6 | 4790 | 76 | ||
XM_021838875.1 | XP_021694567.1 | 5 - 6 | 4787 | 76 | ||
XM_021838876.1 | XP_021694568.1 | 4 - 5 | 2602 | 76 | ||
XM_021838877.1 | XP_021694569.1 | 4 - 5 | 3005 | 76 | ||
XM_021838878.1 | XP_021694570.1 | 4 - 5 | 2593 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
