Assay Details
Gene Symbol: LOC5574316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838876.1 | XP_021694568.1 | 3 - 4 | 2569 | 60 | |
XM_021838877.1 | XP_021694569.1 | 3 - 4 | 2972 | 60 | ||
XM_021838878.1 | XP_021694570.1 | 3 - 4 | 2560 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
