Assay Details
Gene Symbol: LOC5574316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838871.1 | XP_021694563.1 | 21 - 22 | 7784 | 87 | |
XM_021838872.1 | XP_021694564.1 | 20 - 21 | 7776 | 87 | ||
XM_021838874.1 | XP_021694566.1 | 21 - 22 | 7739 | 87 | ||
XM_021838875.1 | XP_021694567.1 | 21 - 22 | 7736 | 87 | ||
XM_021838876.1 | XP_021694568.1 | 20 - 21 | 5551 | 87 | ||
XM_021838877.1 | XP_021694569.1 | 20 - 21 | 5954 | 87 | ||
XM_021838878.1 | XP_021694570.1 | 20 - 21 | 5542 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
