Assay Details
Gene Symbol: LOC5574316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661298.2 | XP_001661348.1 | 16 - 17 | 6945 | 71 | |
XM_021838871.1 | XP_021694563.1 | 17 - 18 | 6952 | 71 | ||
XM_021838872.1 | XP_021694564.1 | 16 - 17 | 6944 | 71 | ||
XM_021838873.1 | XP_021694565.1 | 17 - 18 | 6954 | 71 | ||
XM_021838874.1 | XP_021694566.1 | 17 - 18 | 6907 | 71 | ||
XM_021838875.1 | XP_021694567.1 | 17 - 18 | 6904 | 71 | ||
XM_021838876.1 | XP_021694568.1 | 16 - 17 | 4719 | 71 | ||
XM_021838877.1 | XP_021694569.1 | 16 - 17 | 5122 | 71 | ||
XM_021838878.1 | XP_021694570.1 | 16 - 17 | 4710 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
