Assay Details
Gene Symbol: LOC5572902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847048.1 | XP_021702740.1 | 11 - 12 | 3255 | 103 | |
XM_021847049.1 | XP_021702741.1 | 11 - 12 | 3296 | 103 | ||
XM_021847050.1 | XP_021702742.1 | 11 - 12 | 3080 | 103 | ||
XM_021847051.1 | XP_021702743.1 | 11 - 12 | 3085 | 103 | ||
XM_021847052.1 | XP_021702744.1 | 11 - 12 | 3250 | 103 |

More Information
Set Membership: |
![]() ![]() |
