Assay Details
Gene Symbol: LOC5576280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844448.1 | XP_021700140.1 | 12 - 13 | 2616 | 143 | |
XM_021844450.1 | XP_021700142.1 | 9 - 10 | 1965 | 143 |

More Information
Set Membership: |
![]() ![]() |
