Assay Details
Gene Symbol: LOC5576280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844448.1 | XP_021700140.1 | 13 - 14 | 3339 | 116 | |
XM_021844449.1 | XP_021700141.1 | 12 - 13 | 3365 | 116 | ||
XM_021844450.1 | XP_021700142.1 | 10 - 11 | 2688 | 116 |

More Information
Set Membership: |
![]() ![]() |
