Assay Details
Gene Symbol: LOC5576280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844448.1 | XP_021700140.1 | 14 - 15 | 3394 | 80 | |
XM_021844449.1 | XP_021700141.1 | 13 - 14 | 3420 | 80 | ||
XM_021844450.1 | XP_021700142.1 | 11 - 12 | 2743 | 80 | ||
XM_021844451.1 | XP_021700143.1 | 12 - 13 | 2705 | 80 | ||
XM_021844452.1 | XP_021700144.1 | 12 - 13 | 2703 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
