Assay Details
Gene Symbol: LOC5576280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844448.1 | XP_021700140.1 | 2 - 3 | 285 | 73 | |
XM_021844449.1 | XP_021700141.1 | 1 - 2 | 314 | 73 | ||
XM_021844451.1 | XP_021700143.1 | 1 - 2 | 313 | 73 | ||
XM_021844452.1 | XP_021700144.1 | 1 - 2 | 314 | 73 |
