Assay Details
Gene Symbol: LOC5576280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844448.1 | XP_021700140.1 | 3 - 4 | 505 | 70 | |
XM_021844449.1 | XP_021700141.1 | 2 - 3 | 534 | 70 | ||
XM_021844451.1 | XP_021700143.1 | 2 - 3 | 533 | 70 | ||
XM_021844452.1 | XP_021700144.1 | 2 - 3 | 534 | 70 |

More Information
Set Membership: |
![]() ![]() |
