Assay Details
Gene Symbol: LOC23687451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837701.1 | XP_021693393.1 | 12 - 13 | 3980 | 62 | |
XM_021837702.1 | XP_021693394.1 | 12 - 13 | 3896 | 62 | ||
XM_021837703.1 | XP_021693395.1 | 13 - 14 | 3852 | 62 | ||
XM_021837704.1 | XP_021693396.1 | 13 - 14 | 3843 | 62 | ||
XM_021837705.1 | XP_021693397.1 | 13 - 14 | 3830 | 62 | ||
XM_021837706.1 | XP_021693398.1 | 11 - 12 | 3933 | 62 | ||
XM_021837707.1 | XP_021693399.1 | 10 - 11 | 2284 | 62 | ||
XM_021837708.1 | XP_021693400.1 | 11 - 12 | 3317 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
