Assay Details
Gene Symbol: LOC23687451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837701.1 | XP_021693393.1 | 15 - 16 | 4617 | 89 | |
XM_021837702.1 | XP_021693394.1 | 15 - 16 | 4533 | 89 | ||
XM_021837703.1 | XP_021693395.1 | 16 - 17 | 4489 | 89 | ||
XM_021837704.1 | XP_021693396.1 | 16 - 17 | 4480 | 89 | ||
XM_021837705.1 | XP_021693397.1 | 16 - 17 | 4467 | 89 | ||
XM_021837706.1 | XP_021693398.1 | 14 - 15 | 4570 | 89 | ||
XM_021837707.1 | XP_021693399.1 | 13 - 14 | 2921 | 89 | ||
XM_021837708.1 | XP_021693400.1 | 14 - 15 | 3954 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
