Assay Details
Gene Symbol: LOC23687451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837701.1 | XP_021693393.1 | 2 - 3 | 1182 | 104 | |
XM_021837703.1 | XP_021693395.1 | 3 - 4 | 1055 | 103 | ||
XM_021837704.1 | XP_021693396.1 | 3 - 4 | 1046 | 103 | ||
XM_021837705.1 | XP_021693397.1 | 3 - 4 | 1033 | 103 | ||
XM_021837706.1 | XP_021693398.1 | 2 - 3 | 1178 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021837701.1, XM_021837703.1, XM_021837704.1, XM_021837705.1, XM_021837706.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
