Assay Details
Gene Symbol: LOC23687451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837701.1 | XP_021693393.1 | 3 - 4 | 1845 | 73 | |
XM_021837702.1 | XP_021693394.1 | 3 - 4 | 1761 | 73 | ||
XM_021837703.1 | XP_021693395.1 | 4 - 5 | 1717 | 73 | ||
XM_021837704.1 | XP_021693396.1 | 4 - 5 | 1708 | 73 | ||
XM_021837705.1 | XP_021693397.1 | 4 - 5 | 1695 | 73 | ||
XM_021837706.1 | XP_021693398.1 | 3 - 4 | 1840 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
