Assay Details
Gene Symbol: LOC5576444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841346.1 | XP_021697038.1 | 7 - 8 | 4548 | 75 | |
XM_021841347.1 | XP_021697039.1 | 7 - 8 | 4197 | 75 | ||
XM_021841348.1 | XP_021697040.1 | 6 - 7 | 4543 | 75 | ||
XM_021841349.1 | XP_021697041.1 | 6 - 7 | 3977 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
