Assay Details
Gene Symbol: LOC5576444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841346.1 | XP_021697038.1 | 6 - 7 | 2695 | 104 | |
XM_021841347.1 | XP_021697039.1 | 6 - 7 | 2344 | 104 | ||
XM_021841348.1 | XP_021697040.1 | 5 - 6 | 2690 | 104 | ||
XM_021841349.1 | XP_021697041.1 | 5 - 6 | 2124 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
