Assay Details
Gene Symbol: LOC5576444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841346.1 | XP_021697038.1 | 1 - 2 | 908 | 95 | |
XM_021841347.1 | XP_021697039.1 | 1 - 2 | 937 | 95 | ||
XM_021841348.1 | XP_021697040.1 | 1 - 2 | 908 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021841346.1, XM_021841347.1, XM_021841348.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
