Assay Details
Gene Symbol: LOC5569782 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658670.2 | XP_001658720.2 | 3 - 4 | 1137 | 68 | |
XM_021855721.1 | XP_021711413.1 | 3 - 4 | 1369 | 68 | ||
XM_021855722.1 | XP_021711414.1 | 4 - 5 | 2228 | 68 | ||
XM_021855723.1 | XP_021711415.1 | 4 - 5 | 1010 | 68 | ||
XM_021855724.1 | XP_021711416.1 | 4 - 5 | 1063 | 68 | ||
XM_021855725.1 | XP_021711417.1 | 4 - 5 | 1115 | 68 | ||
XM_021855726.1 | XP_021711418.1 | 4 - 5 | 1083 | 68 | ||
XM_021855727.1 | XP_021711419.1 | 4 - 5 | 2376 | 68 |

More Information
Set Membership: |
![]() ![]() |
