Assay Details
Gene Symbol: LOC5569782 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658670.2 | XP_001658720.2 | 4 - 5 | 1424 | 110 | |
XM_021855721.1 | XP_021711413.1 | 4 - 5 | 1656 | 110 | ||
XM_021855722.1 | XP_021711414.1 | 5 - 6 | 2515 | 110 | ||
XM_021855723.1 | XP_021711415.1 | 5 - 6 | 1297 | 110 | ||
XM_021855724.1 | XP_021711416.1 | 5 - 6 | 1350 | 110 | ||
XM_021855725.1 | XP_021711417.1 | 5 - 6 | 1402 | 110 | ||
XM_021855726.1 | XP_021711418.1 | 5 - 6 | 1370 | 110 | ||
XM_021855727.1 | XP_021711419.1 | 5 - 6 | 2663 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
