Assay Details
Gene Symbol: LOC5578606 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842248.1 | XP_021697940.1 | 2 - 3 | 2064 | 62 | |
XM_021842249.1 | XP_021697941.1 | 3 - 4 | 1983 | 62 | ||
XM_021842250.1 | XP_021697942.1 | 3 - 4 | 1843 | 62 |

More Information
Set Membership: |
![]() ![]() |
