Assay Details
Gene Symbol: LOC5575651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838070.1 | XP_021693762.1 | 1 - 2 | 796 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021838070.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
