Assay Details
Gene Symbol: LOC5579338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848420.1 | XP_021704112.1 | 2 - 3 | 688 | 78 | |
XM_021848421.1 | XP_021704113.1 | 2 - 3 | 622 | 78 | ||
XM_021848422.1 | XP_021704114.1 | 2 - 3 | 688 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
