Assay Details
Gene Symbol: LOC5579338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848420.1 | XP_021704112.1 | 8 - 9 | 2041 | 107 | |
XM_021848421.1 | XP_021704113.1 | 8 - 9 | 1975 | 107 | ||
XM_021848422.1 | XP_021704114.1 | 7 - 8 | 2020 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
