Assay Details
Gene Symbol: LOC5563715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837535.1 | XP_021693227.1 | 1 - 2 | 2911 | 86 | |
XM_021837536.1 | XP_021693228.1 | 1 - 2 | 2231 | 86 | ||
XM_021837537.1 | XP_021693229.1 | 1 - 2 | 2913 | 86 |
