Assay Details
Gene Symbol: LOC5563715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837535.1 | XP_021693227.1 | 7 - 8 | 3624 | 80 | |
XM_021837536.1 | XP_021693228.1 | 6 - 7 | 2884 | 80 | ||
XM_021837537.1 | XP_021693229.1 | 7 - 8 | 3626 | 80 |
