Assay Details
Gene Symbol: LOC5563715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837535.1 | XP_021693227.1 | 9 - 10 | 3895 | 71 | |
XM_021837536.1 | XP_021693228.1 | 8 - 9 | 3155 | 71 |
