Assay Details
Gene Symbol: LOC5580104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848697.1 | XP_021704389.1 | 4 - 5 | 3573 | 97 | |
XM_021848704.1 | XP_021704396.1 | 4 - 5 | 3579 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
