Assay Details
Gene Symbol: LOC5576538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839109.1 | XP_021694801.1 | 1 - 2 | 1005 | 193 | |
XM_021839110.1 | XP_021694802.1 | 2 - 3 | 750 | 193 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021839109.1, XM_021839110.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
