Assay Details
Gene Symbol: LOC5576538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839109.1 | XP_021694801.1 | 11 - 12 | 7621 | 89 | |
XM_021839110.1 | XP_021694802.1 | 12 - 13 | 7366 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
