Assay Details
Gene Symbol: LOC5571076 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659467.2 | XP_001659517.1 | 4 - 5 | 1993 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5571076 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659467.2 | XP_001659517.1 | 4 - 5 | 1993 | 79 |
Set Membership: |
![]() ![]() ![]() ![]() |