Assay Details
Gene Symbol: LOC5574949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655366.2 | XP_001655416.2 | 2 | 520 | 67 | |
XM_021852979.1 | XP_021708671.1 | 2 | 524 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
