Assay Details
Gene Symbol: LOC5567983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842011.1 | XP_021697703.1 | 2 - 3 | 94 | ||
XM_021842012.1 | XP_021697704.1 | 2 - 3 | 94 | |||
XM_021842013.1 | XP_021697705.1 | 2 - 3 | 94 | |||
XM_021842014.1 | XP_021697706.1 | 2 - 3 | 94 | |||
XM_021842015.1 | XP_021697707.1 | 2 - 3 | 94 | |||
GenBank mRNA | KF227903.1 | - | 2 - 3 | 127 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021842011.1, XM_021842012.1, XM_021842013.1, XM_021842014.1, XM_021842015.1 transcripts.
Set Membership: |
![]() ![]() |
