Assay Details
Gene Symbol: LOC5567983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842011.1 | XP_021697703.1 | 3 - 4 | 236 | 79 | |
XM_021842012.1 | XP_021697704.1 | 3 - 4 | 191 | 79 | ||
XM_021842013.1 | XP_021697705.1 | 3 - 4 | 160 | 79 | ||
XM_021842014.1 | XP_021697706.1 | 3 - 4 | 237 | 79 | ||
XM_021842015.1 | XP_021697707.1 | 3 - 4 | 195 | 79 | ||
GenBank mRNA | KF227903.1 | - | 3 - 4 | 166 | 79 |
