Assay Details
Gene Symbol: LOC5567983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842011.1 | XP_021697703.1 | 4 - 5 | 300 | 91 | |
XM_021842012.1 | XP_021697704.1 | 4 - 5 | 255 | 91 | ||
XM_021842013.1 | XP_021697705.1 | 4 - 5 | 224 | 91 | ||
XM_021842014.1 | XP_021697706.1 | 4 - 5 | 301 | 91 | ||
XM_021842015.1 | XP_021697707.1 | 4 - 5 | 259 | 91 | ||
GenBank mRNA | KF227903.1 | - | 4 - 5 | 230 | 91 |
