Assay Details
Gene Symbol: LOC5567983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842011.1 | XP_021697703.1 | 6 - 7 | 101 | ||
XM_021842012.1 | XP_021697704.1 | 6 - 7 | 101 | |||
XM_021842013.1 | XP_021697705.1 | 6 - 7 | 101 | |||
GenBank mRNA | KF227903.1 | - | 6 - 7 | 797 | 101 |

More Information
Set Membership: |
![]() ![]() |
