Assay Details
Gene Symbol: LOC5577291 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656342.2 | XP_001656392.1 | 3 | 1236 | 67 | |
GenBank mRNA | AY432865.1 | - | 1 | 519 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
