Assay Details
Gene Symbol: LOC5564792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849190.1 | XP_021704882.1 | 2 - 3 | 933 | 71 | |
GenBank mRNA | AY433484.1 | - | 1 - 2 | 334 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
