Assay Details
Gene Symbol: LOC5572646 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654095.2 | XP_001654145.2 | 4 - 5 | 3803 | 91 | |
XM_021845115.1 | XP_021700807.1 | 5 - 6 | 3932 | 91 | ||
GenBank mRNA | AY433562.1 | - | 2 - 3 | 441 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
