Assay Details
Gene Symbol: LOC23687643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837683.1 | XP_021693375.1 | 2 - 3 | 437 | 60 | |
GenBank mRNA | AY433568.1 | - | 2 - 3 | 402 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
