Assay Details
Gene Symbol: LOC5567494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657439.2 | XP_001657489.1 | 1 - 2 | 231 | 66 | |
GenBank mRNA | AY433602.1 | - | 1 - 2 | 216 | 66 | |
DQ440112.1 | - | 1 - 2 | 98 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
