Assay Details
Gene Symbol: LOC5573557 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848064.1 | XP_021703756.1 | 3 | 1706 | 74 | |
GenBank mRNA | AY433737.1 | - | 1 | 522 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
