Assay Details
Gene Symbol: LOC5572285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653920.2 | XP_001653970.1 | 1 - 2 | 290 | 78 | |
GenBank mRNA | AY433757.1 | - | 1 - 2 | 56 | 78 | |
DQ440263.1 | - | 1 - 2 | 67 | 78 |
