Assay Details
Gene Symbol: LOC5569538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652800.2 | XP_001652850.2 | 2 - 3 | 1319 | 82 | |
XM_021851080.1 | XP_021706772.1 | 2 - 3 | 1319 | 82 | ||
GenBank mRNA | AY438004.1 | - | 2 - 3 | 1289 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
